Thermal transfer ribbon for printer, R7964 series
Lieferant: Brady
237588
BRDY237588EA 41.97 EUR
BRDY237588
Thermal transfer ribbon for printer, R7964 series
Farbbänder
If the R-7964 print ribbon is printed on the correct Brady label, this guarantees good solvent resistance and razor sharp print quality.
Verpackung: One print ribbon/box.