Brady® FR22 RFID Fixed Reader
Lieferant: Brady
311937
BRDY311937EA 1928.44 EUR
BRDY311937
Brady® FR22 RFID Fixed Reader
Barcode- und RFID-Systeme
The BRADY FR22 is the core device of a new approach for RFID fixed systems with wide communication options and a modular design. The FR22 is a critical building block for various track and trace and intelligent manufacturing solutions. It enables different use cases for different segments depending on which modules are attached to it.
  • Fast and reliable edge computing with support for custom embedded applications
  • New UHF RFID engine based on the very latest technology
  • Brady extension connector for plug and play accessories like Brady MUX16
  • Connect external devices, sensors, triggers, lights and buzzers
  • Full set of API’s available for controlling the reader
Wide communication options that enable flexibility in system designing and no need for external connectivity hardware. Develop embedded web apps with user interface. Eliminating the need for external PC/tablet.
Standards: ISO 18000-63 (EPC Class 1Gen2v2).
Bestellinformation: Two years warranty (extendable), Free technical support during and after warranty. This box includes a Brady FR22 fixed reader, a WLAN/Bluetooth antenna, a safety and regulations card, additional product label with product information.