Polyethylene Tags for M710, M610 and M611, Bulk
Lieferant: Brady
312126
BRDY312126EA 232.26 EUR
BRDY312126
Polyethylene Tags for M710, M610 and M611, Bulk
Etiketten
Multipurpose tags can be used for a variety of applications including identification of multi-conductor cables, inventory, equipment, lockout, safety warning repair and work-in-progress. Extremely tear-resistant and superior cold-weather tag.
  • Cross-laminated polyethylene provides extreme tear resistance and excellent cold weather performance
  • Matte finish
Applications include data and telecommunications labelling, wire and cable labelling, inventory and inspection labelling, lockout tagout, and safety identification.
Verpackung: 1000 tags per box.