Matte White Polypropylene RFID Laboratory Labels
Lieferant: Brady
312403
BRDY312403EA 231.41 EUR
BRDY312403
Matte White Polypropylene RFID Laboratory Labels
Etiketten
UHF RFID label for use on general laboratory identification on medical well plate, petri and cell culture dish.
  • Pharmaceutical and healthcare
  • The RFID label is designed for used on well plate, petri and cell culture dish in the laboratory environment
  • Polyester tag face material, with chip bonded to aluminium antenna using anisotropic conductive film adhesive, suitable for thermal transfer printing, including RFID encoding
  • Labels are outside wound and 100% inspected, 500 good RFID labels per roll with bad tags marked
Standards: Compliant with directive 2011/65/EU and 2015/863.