Label Printer, QWERTY EU Wifi, Bluetooth + BWS SFID Suite, M710
Lieferant: Brady
317830
BRDY317830EA 1229 EUR
BRDY317830
Label Printer, QWERTY EU Wifi, Bluetooth + BWS SFID Suite, M710
Drucker Label Printers
Portable printer.
  • Stay connected wherever your work happens with enhanced Bluetooth and Wi-Fi
  • Get lightning-fast print speeds: 76,2 mm per second, plus cut labels automatically
  • Work longer – print 4500 labels on a full battery charge
  • Enjoy more durability with a printer that withstands 1,20 m drops, blowing sand and dust
  • Get more work done using a keypad or software to print labels
  • Print what you need, how you need – text, barcodes, shapes and images using a keypad or software
  • Get shop-to-field flexibility with label saving, sharing and data imports
  • This
More Product Information