Label Printer, EU, M611
Lieferant: Brady
317844
BRDY317844EA 595 EUR
BRDY317844
Label Printer, EU, M611
Drucker Label Printers
Portable printer.
  • Create and print up to 50,80 mm width labels
  • Print labels at 50,80 mm per second print speed
  • Design, preview and print any way you choose: from your phone, tablet, or computer
  • Store more than 1000 labels on the printer, then print at the jobsite
  • Create and share label files to print from anywhere using Bluetooth or Wi-Fi
  • Features a compact and lightweight design
  • Allows connectivity for up to 5 users via Bluetooth
Get more flexibility in the field. Design. Preview. Print. Any way you choose. Store more than 1000 labels on board, access files easier and print multiple ways – on demand. Modernise workflows: create labels in the office and share to the field to print anywhere, anytime.
Standards: CE 2014/53/EU RED, 2014/35/EU LVD, 2014/30/EU EMCD, 2011/65/EU, RoHS, UL 60950-1 2nd Edition, EN \ IEC 60950-1:2006 + A11:2009 + A1:2010 + A12:2011+ A2:2013, EN \ IEC 62368-1:2014, WEEE.
Zubehörinformationen: M6-;M6C-;BM-;BMC-.
Lieferumfang: This printer ships in a cardboard box holding a USB cable, a quick start guide, an AC powercord, a USB stick holding product info and Windows driver, a Brady Workstation Basic Design Suite, a roll of M6-11-427 and an M61-R4310 ribbon.