Magicard™ MA1000K Monochromfilme
Lieferant: Brady
321959
BRDY321959EA 18 EUR
BRDY321959
Magicard™ MA1000K Monochromfilme
Farbbänder
The Magicard™ MA1000K black ribbon is perfect for adding contrasting prints.
  • High-quality, monochrome ribbon guarantees to print 1000 single-sided cards
  • Includes a cleaning roller for dust-free printing
Applications: Healthcare and hospital labelling, facility identification, security identification, general identification.
Lieferumfang: 1 dye film + 1 cleaning roller.