Magicard™ Monochrome Film, Gold
Lieferant: Brady
321961
BRDY321961EA 75 EUR
BRDY321961
Magicard™ Monochrome Film, Gold
Farbbänder
The Magicard™ MA1000K gold ribbon is perfect for adding gold accents to prints.
  • High-quality, monochrome ribbon guarantees to print 1000 single-sided cards
  • Includes a cleaning roller for dust-free printing
Applications: Healthcare and hospital labelling, facility identification, security identification, general identification.
Lieferumfang: 1 dye film + 1 cleaning roller.