Magicard™ Colour Film for Magicard™ 600, 300 Shot
Lieferant: Brady
321969
BRDY321969EA 70 EUR
BRDY321969
Magicard™ Colour Film for Magicard™ 600, 300 Shot
Farbbänder
The Magicard™ colour ribbon is perfect for full colour prints with a Magicard™ 600 printer.
  • High-quality, full colour ribbon
  • Yellow, Magenta, Cyan, Black with an overcoat
  • Includes a cleaning roller for dust-free printing
Print 300 single-sided cards.
Lieferumfang: Includes 1 dye film and 1 cleaning roller.