Magicard™ Prima 8 Re-Transfer Film
Lieferant: Brady
321980
BRDY321980EA 115 EUR
BRDY321980
Magicard™ Prima 8 Re-Transfer Film
Farbbänder
The Magicard™ Prima 8 transfer film allows users to transfer colours on the cards.
  • Allows for over-the-edge printing
  • Guarantees to print 1000 single-sided cards
Transfer colours on the cards.
Lieferumfang: 1 re-transfer film.