Self-Laminating Vinyl Wire and Cable Labels for Wraptor™ A6500 Printer
Lieferant: Brady
801616
BRDY801616EA 376.5 EUR
BRDY801616
Self-Laminating Vinyl Wire and Cable Labels for Wraptor™ A6500 Printer
Etiketten
Self-laminating vinyl labels for the Brady Wraptor™ A6500 Wire ID Printer Applicator. For smooth and textured or rough surfaces.
  • Excellent resistance to water, oil and solvents
  • Matt finish
  • R4300 ribbon
Ideal for data and telecommunications labelling, electrical labelling, laboratory labelling, wire and cable labelling.
Standards: This material is UL recognised with its respective ribbon