Economy breaker lockout, multi-pole
Lieferant: Brady
805814
BRDY805814EA 15.66 EUR
BRDY805814
Economy breaker lockout, multi-pole
Sicherheitsverriegelungen
This lockout is designed to fit multi-pole breakers.
  • Made from impact resistant nylon
  • One padlock can be applied
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm