Portable padlock station
Lieferant: Brady
811218
BRDY811218EA 122.83 EUR
BRDY811218
Portable padlock station
Sicherheitsverriegelungen
This portable storage station ensures that lockout devices remain easily accessible and ready for use whenever needed.
  • Made from rugged and rust-proof PP
  • Attachable handle for easy portability
  • Screw slots for permanent wall mounting
  • Lockable, semi-transparent cover protects the contents from exposure to harsh environments
Bestellinformation: Station is supplied empty (without contents).