Oil drum top covers
Lieferant: Brady
813784
BRDY813784EA 142.1 EUR
BRDY813784
Oil drum top covers
Absorptionsmaterial
Oil sorbent drum top cover with pre-cut holes that absorbs oil and petroleum based fluids.
This helps to keep drum top, work floor and surrounding storage area neat and clean by preventing drips from reaching the floor.