Universal sorbent pads
Lieferant: Brady
813809
BRDY813809EA 122.77 EUR
BRDY813809
Universal sorbent pads
Absorptionsmaterial
These grey coloured heavy weight, bonded and perforated universal sorbent pad absorbs industrial liquids including oils, water, solvents, coolants.
  • One sided spunbound for extra strength and less lint
  • Made of two-ply, spunbound/meltblown, bonded
Perforated to help minimise waste and lower disposal costs. Ideal sorbent for dealing with leaks and drips in the workplace and also suited for general industrial maintenance and spill clean up.