Chemical sorbent socs
Lieferant: Brady
813850
BRDY813850EA 115.58 EUR
BRDY813850
Chemical sorbent socs
Absorptionsmaterial
These chemical sorbent socs filled with chemical meltblown polypropylene that absorbs chemicals and hazardous liquids.
  • Strong wicking action
  • Green colour
  • Ideal sorbent for containing and absorbing leaks and spills
Absorption begins instantly after contact with the spilled liquid, no need to turn the soc. Flexible design allows the soc to fit around any machine, corner or curve.