Chemical Drum Spill Kit
Lieferant: Brady
813877
BRDY813877EA 1263.92 EUR
BRDY813877
Chemical Drum Spill Kit
Absorptionsmaterial
The drum spill kit is designed for large spills of chemicals and hazardous liquids.
  • Drum spill kit that comes in a polyethylene, screw top container
  • Water and chemical resistant to protect the sorbent material
  • Drum can also be used as shipping container for used sorbents
  • Moulded design allows for easy gripping and pick up by a forklift or trolley
Lieferumfang: Supplied with 100 pads (410×510 mm), 8 socs (Ø 7,6×3660 mm), 12 socs (Ø 7,6×1220 mm), 8 pillows (430×480 mm), 50 wipes (300×380 mm), 1 pair of gloves, 1 pair of goggles and 10 disposal bags.