Standard mantory signs
Lieferant: Brady
818421 819165 818865 819464 819465 819315 819316 819017 819018 819166 819167 818422 820060 820061 819762 819763 818272 820358 818273 821699 821700 820359 820209 820210 818123 818124 819613 819614 821401 821402 821103 814163 814164 819016 818420 819761 818423 819019 819314 818418 818280 818429 818415 818417 818430 818271 818276 818277 818278 818419 818279 818867 818116 818117
116-0516EA 4.22 EUR
116-0516 BRDY819165 BRDY818865 116-0508 116-0509 116-0510 116-0511 116-0512 116-0513 116-0514 116-0515 116-0517 116-0518 116-0519 116-0520 116-0521 116-0522 116-0526 116-0523 116-0524 116-0525 116-0527 116-0528 116-0529 116-0530 116-0531 116-0534 116-0535 116-0536 116-0537 116-0540 116-0542 116-0543 BRDY819016 BRDY818420 BRDY819761 BRDY818423 BRDY819019 BRDY819314 BRDY818418 BRDY818280 BRDY818429 BRDY818415 BRDY818417 BRDY818430 BRDY818271 BRDY818276 BRDY818277 BRDY818278 BRDY818419 BRDY818279 BRDY818867 BRDY818116 BRDY818117
Standard mantory signs
Warn- und Verbotszeichen
Mantory signs have always a blue circular backround which contains a white pictogram.These signs display a message or action that must be carried out in a designated area.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm
  • Different sizes
Bestellen
TECHNISCHE DATEN
Environmentally Preferable