Glow-in-the-dark Safety Sign
Lieferant: Brady
836146
BRDY836146EA 6.52 EUR
BRDY836146
Glow-in-the-dark Safety Sign
Warn- und Verbotszeichen
Rigid polypropylene laminated with a printed flexible photoluminescent polyester tape Class B (Nite­Glo B­7568).
  • Substrate type: Rigid photoluminescent polypropylene sign
  • Standard legend colours: Black, blue, green and red
  • Triangle shape; glose finish
  • Maximum service temperature: 66 °C
  • Minimum service temperature: −40 °C
  • Thickness: 1,77 mm
B­7570 is used for safety applications such as safety signage in building, marine, rail and military applications.
Standards: Luminous compliance according DIN 67510, ASTM 2072, ISO 15370, EU marine equipment directive, IMO RES A. 752 (18),
PSPA Class B.