Universal fuse lockout device
Lieferant: Brady
873367
BRDY873367EA 15.66 EUR
BRDY873367
Universal fuse lockout device
Sicherheitsverriegelungen
This fuse lockout device has been designed to prevent the unintended re-energisation of an energy source through a fuse. The device fits most fuse holders between 20 Amps and 400 Amps.
  • Made of nylon with ZP metal slotted set screw
  • With legend 'Danger Do Not Remove Locked Out'
  • Easy to use
Instructions: Clamp onto the fuse holder and use a small flathead screwdriver to tighten the set screw in the device. Place a padlock through the hole to prevent access to the set screw.