Safety lockouts, dual level
Lieferant: Brady
852439 873384
BRDY852439EA 52.91 EUR
BRDY852439 BRDY873384
Safety lockouts, dual level
Sicherheitsverriegelungen
Especially designed to allow for the removal of personal padlocks these lockouts enable a supervisory padlock (or two padlocks on the 12 holes) to remain safely locked in place, ensuring that the machine remains isolated.
  • Foolproof access to the hasp entry and a tamperproof locking facility adds another degree of safety
  • Manufactured from marine grade 316 stainless steel
  • 5 mm shackle will fit most electrical switch gear, cabinets and distribution boards
Shackle W×D×H: 35×5×35 mm
Bestellinformation: Available in 6 hole×1 supervisory, or 12 holes×2 supervisory configurations.