Labels, pipe marking
Lieferant: Brady
N002370 N004112 N004319 N004483 N004511 N004522 N002355 N000485 N003331 N003252 N003298 N005130 N005108 N005163 N004915
BRDYN002370EA 16.12 EUR
BRDYN002370 BRDYN004112 BRDYN004319 BRDYN004483 BRDYN004511 BRDYN004522 BRDYN002355 BRDYN000485 BRDYN003331 BRDYN003252 BRDYN003298 BRDYN005130 BRDYN005108 BRDYN005163 BRDYN004915
Labels, pipe marking
Etiketten
These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.
  • Promotes safety and simplifies maintenance
  • Arrows are pre-cut, and hence does not requires scissors additionally
  • Resistant to temperatures from –40 to +120 °C
  • Resistant to ultraviolet rays and conditions
The markers have a frame that contrasts with the color of the text to increase visibility.