Pipe marker, H₂S
Lieferant: Brady
N004308
BRDYN004308EA 12.27 EUR
BRDYN004308
Pipe marker, H₂S
Warn- und Verbotszeichen
Individual pipe markers on map with perforated arrows, with pictogram.
  • Rubber-based permanent adhesive
  • Polyester with polyester overlaminate material
  • Colour: Black on yellow