Pipe Marker, Disposal
Lieferant: Brady
N005738
BRDYN005738EA 12.27 EUR
BRDYN005738
Pipe Marker, Disposal
Warn- und Verbotszeichen
Individual pipe markers on map with perforated arrows, without pictogram - drain.
  • Polyester with polyester overlaminate
  • Permanent rubber-based adhesive
  • Colour: White on green
Verpackung: Total quantity per pack - five marker(s)/card.