SPC® Oil-only Absorbents
Lieferant: Brady
SKO-1100
BRDYSKO-1100EA 0 EUR
BRDYSKO-1100
SPC® Oil-only Absorbents
Absorptionsmaterial
Drop front spill kit designed for larger spills of oil and petroleum based liquids. Top and drop front lid provides easy access to the absorbents inside the kit.
  • Absorbs up to 972 litres (full content of an IBC)
  • Absorbents and spill control
  • Drop front for fast and easy access
Wheeled to easily manoeuvre to location of the spill.
Lieferumfang: Includes: 300 ENV200-M pads, 2 OP15-E rolls, 4 ENV510 booms, 30 OIL430 socs, 10 OIL 124 socs, 20 OIL1818 pillows, 1 pair SPK-HANDSCH gloves, 1 pair SPK-BRIL goggles, 50 disposal bags.