Handheld microscope, BMS Trekker
Lieferant: BREUKHOVEN
76520
BREU76520EA 0 EUR
BREU76520
Handheld microscope, BMS Trekker
Mikroskope
The BMS Trekker is a high quality handheld microscope that enhances objects up to 35 times.
  • Detachable eyepiece to be used as a 10× magnification latch
  • Built-in white LED with daylight correction – efficient energy saving lamp
  • Can be connected to any SLR camera
Suitable for outdoor and indoor use in technology, science, schools and leisure.