Compact automatic burettes, Schellbach, BLAUBRAND®
Lieferant: Brand
23872
BRND23872EA 164 EUR
BRND23872
Compact automatic burettes, Schellbach, BLAUBRAND®
Büretten
Borosilicate glass 3.3, class AS, DE-M marking.
Lieferumfang: Burettes supplied with 1000 ml PE bottle.