Compact burettes, interchangeable PTFE stopcock, BLAUBRAND®
Lieferant: Brand
23924 23923 23925
BRND23924EA 94.2 EUR
BRND23924 BRND23923 BRND23925
Compact burettes, interchangeable PTFE stopcock, BLAUBRAND®
Büretten
Borosilicate 3.3 glass, class AS.
  • Calibrated to deliver ( TD, Ex )
  • Schellbach stripe for accurate meniscus reading
  • Interchangeable PTFE stopcock
  • High contrast blue enamel graduations
  • Batch certificate included
Modular automatic burette concept. Fast to dismantle and easy to clean. Easy to repair - all individual components are replaceable.
All BLAUBRAND® automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging. On request, they are also available with an individual certificate, USP individual certificate or DAkkS calibration certificate.
Standards: In compliance with DIN EN ISO 385 standards.