Compact automatic burettes, amber, Schellbach, BLAUBRAND®
Lieferant: Brand
23939
BRND23939EA 200 EUR
BRND23939
Compact automatic burettes, amber, Schellbach, BLAUBRAND®
Büretten
Borosilicate glass 3.3 bottle, class AS, DE-M marking.
  • Modular concept - all individual components are replaceable
  • Fast to dismantle and easy to clean and repair
  • Calibrated to deliver 'TD, Ex'
  • Detachable PTFE stopcock
  • Automatic zeroing and Schellbach stripe
Standards: DIN EN ISO 385
Bestellinformation: Please order burette clamps, supports and rubber bellows separately.
Lieferumfang: Supplied as burette tube, stopcock with precision tip, filling tube (PVC, transparent), pump head and 2000 ml bottle.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable