Schülke Wipes 70
Lieferant: Brand
44870
BRND44870EA 155 EUR
BRND44870
Schülke Wipes 70
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
Fleece wipes for impregnation with universal or disinfecting detergents.
  • Made of 100% polyester, dry
The lint-free and tear-proof wipes are designed for both optimal disinfectant delivery and excellent contaminant uptake. Due to their size they are particularly suitable to disinfect your workplace, lab table or clean bench quickly and reliably.
The wipes can be stored in the convenient dispenser box.
For high-volume use, wipes can be soaked with liquid disinfectant directly in the handy dispenser box (lasts up to 28 days).
Works well with the surface disinfecting detergent Pursept® A Xpress.