Buckets
Lieferant: Brand
71772 71774
BRND71772EA 17.5 EUR
BRND71772 BRND71774
Buckets
Eimer
HDPE, white.
  • Good chemical and temperature resistance
  • Scale with 1 litre steps
  • Without spout and lid (Please order push-on lid seperately)
For industrial and laboratory applications. With handle.