Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery (individual certificate)
Lieferant: Brand
927722 927725 927727 927729 927731
BRND927722EA 30.6 EUR
BRND927722 BRND927725 BRND927727 BRND927729 BRND927731
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery (individual certificate)
Pipetten Messpipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 1: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • Individual certificate
  • Only pipettes marked (*) are suitable for cotton plugs
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Standards: DIN EN ISO 835. DE-M marking.