Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery, with DAkkS certificate
Lieferant: Brand
DAKKS27727 DAKKS27729 DAKKS27731 DAKKS27721 DAKKS27722 DAKKS27724
BRNDDAKKS27727EA 77.7 EUR
BRNDDAKKS27727 BRNDDAKKS27729 BRNDDAKKS27731 BRNDDAKKS27721 BRNDDAKKS27722 BRNDDAKKS27724
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery, with DAkkS certificate
Pipetten Messpipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 1: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • DAkks calibration certificate
  • Only pipettes marked (*) are suitable for cotton plugs
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Standards: DIN EN ISO 835. DE-M marking.