Bio RS-24 Mini-Rotator
Lieferant: BIOSAN
BS-010133-AAG
BSANBS-010133-AAGEA 518 EUR
BSANBS-010133-AAG
Bio RS-24 Mini-Rotator
Schüttler Rotations-/Wippschüttler
Mini-rotator Bio RS-24 provides vertical rotation of the platform. The rotator is an ideal instrument for preventing blood coagulation in tubesand for fulfilment of procedures of biological components extraction.
  • Digital time setting: 1 min to 24 h/non-stop (increment 1 min)
  • Vertical rotation movement: Overhead, 360°
  • External power supply: Input AC 100 to 240 V; 50/60 Hz; Output DC 12 V
The device is simple to operate it is designed as a low cost solution. Rotator can be used in cold rooms or incubators, operating at ambient temperature range 4 to 40 °C.