Anti-CD163 Polyclonal Antibody

Lieferant: BosterBio
A00812-1
BSBTA00812-1EA 552 EUR
BSBTA00812-1
Anti-CD163 Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CD163 detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CD163
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR