Anti-TBR2/Eomes Polyclonal Antibody

Lieferant: BosterBio
A00992-1
BSBTA00992-1EA 552 EUR
BSBTA00992-1
Anti-TBR2/Eomes Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for TBR2/Eomes detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: TBR2/Eomes
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR