PicoKine Mouse IL-17D; ELISA Kit

Lieferant: Boster Bio

EK0792
BSBTEK0792EA 646 EUR
BSBTEK0792
PicoKine Mouse IL-17D; ELISA Kit
Tests ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of mouse IL-17D.


  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect mouse IL17D with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. IL17D tissue specificity:


96wells/kit, with removable strips.

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Target protein IL-17D
Reaktivität der primären Antikörper Mouse
Testtyp Sandwich
Wirt

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR