PicoKine Human MBL2/MPB-C; ELISA Kit
Lieferant: Boster Bio
Sandwich high sensitivity ELISA kit for quantitative detection of human MBP-C/MBL2
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect human Mbl/MBL2 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Mbl/MBL2 tissue specificity: Plasma protein produced mainly in the liver.
96wells/kit, with removable strips.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...