PicoKine Human Caspase 3; ELISA Kit

Lieferant: BosterBio

EK1425
BSBTEK1425EA 646 EUR
BSBTEK1425
PicoKine Human Caspase 3; ELISA Kit
Tests ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of human Caspase 3.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect Human Caspase 3/CASP3 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernate sand serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Caspase 3/CASP3 tissue specificity: Highly expressed in lung, spleen, heart, liver and kidney. Moderate levels in brain and skeletal muscle, and low in testis. Also found in many cell lines, highest expression in cells of the immune system.


96wells/kit, with removable strips.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR