PicoKine Mouse PD-L1/B7-H1; ELISA Kit
Lieferant: BosterBio
Sandwich high sensitivity ELISA kit for quantitative detection of mouse PD-L1/B7-H1.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect Mouse Pd L1/Cd274 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Pd L1/Cd274 tissue specificity: Highly expressed in the heart, thymus, skeletal muscle, and lung. Weakly expressed in the kidney, spleen, thyroid, and liver. Expressed on activated dendritic cells, B- cells and macrophages. Expressed in numerous tumor cells lines of lymphoid origin. .
96wells/kit, with removable strips.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...