PicoKine Mouse IDS/Iduronate 2 Sulfatase; ELISA Kit

Lieferant: Boster Bio

EK1453
BSBTEK1453EA 646 EUR
BSBTEK1453
PicoKine Mouse IDS/Iduronate 2 Sulfatase; ELISA Kit
Tests ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of mouse IDS.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect Mouse Iduronate 2-Sulfatase/Ids with <15 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Iduronate 2-Sulfatase/Ids tissue specificity: Found to be expressed in alpha and beta pancreatic cells. .


96wells/kit, with removable strips.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR