PicoKine Human APOA1/Apoa I; ELISA Kit
Lieferant: Boster Bio
Sandwich high sensitivity ELISA kit for quantitative detection of human APOA1.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect Human Apoa I/APOA1 with <50 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(heparin, EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Apoa I/APOA1 tissue specificity: Major protein of plasma HDL, also found in chylomicrons. Synthesised in the liver and small intestine. The oxidized form at Met-110 and Met-136 is increased in individuals with increased risk for coronary artery disease, such as in carrier of the eNOSa/b genotype and exposure to cigarette smoking. It is also present in increased levels in aortic lesions relative to native ApoA-I and increased levels are seen with increasing severity of disease. .
96wells/kit, with removable strips.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...