Anti-VWF Polyclonal Antibody

Lieferant: BosterBio
PB9062
BSBTPB9062EA 552 EUR
BSBTPB9062
Anti-VWF Polyclonal Antibody
Antikörper
Polyclonal antibody for VON WILLEBRAND FACTOR/VWF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. VON WILLEBRAND FACTOR/VWF information: Molecular Weight: 309265 MW; Subcellular Localization: Secreted . Secreted, extracellular space, extracellular matrix . Localized to storage granules; Tissue Specificity: Plasma.

Type: Primary
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR