Anti-CYP2E1 Polyclonal Antibody

Lieferant: BosterBio
PB9190
BSBTPB9190EA 552 EUR
BSBTPB9190
Anti-CYP2E1 Polyclonal Antibody
Antikörper
Polyclonal antibody for CYP2E1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. CYP2E1 information: Molecular Weight: 56849 MW; Subcellular Localization: Endoplasmic reticulum membrane; Peripheral membrane protein. Microsome membrane; Peripheral membrane protein.

Type: Primary
Antigen: CYP2E1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR