proBDNF Rapid™ ELISA Kit: Human, Mouse, Rat
Lieferant: Biosensis
BEK-2217-1P BEK-2217-2P
BSENBEK-2217-1PEA 398 EUR
BSENBEK-2217-1P BSENBEK-2217-2P
proBDNF Rapid™ ELISA Kit: Human, Mouse, Rat
Tests ELISAs
The Biosensis proBDNF Rapid™ enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the specific, fast and reliable quantification of proBDNF in less than 4 hours in cell culture supernatants, cell lysates, serum, citrate-plasma and tissue extracts only if used as directed.
This proBDNF Rapid™ ELISA Kit employs a recombinant, cleavage-resistant human proBDNF standard produced by Biosensis and validated against externally available proBDNF proteins.Due to a high degree of amino acid sequence homology, mouse and rat proBDNF can be quantified and expressed as human proBDNF equivalents. Internal Biosensis validation suggests that the use of the human standard provided in this kit will provide estimates that are identical, or close, to the actual levels of rat and mouse proBDNF present in rodent samples.
The purpose of this kit is the in vitro quantitative determination of human proBDNF in samples such as serum, plasma (EDTA and citrate) and cell culture supernatants.
Note that accurate proBDNF quantification in human serum and citrate-plasma requires the addition of Heterophilic Antibody Blocker provided in the kit, and available for purchase separately.
This kit consists of a pre-coated polyclonal anti-proBDNF capture antibody, a biotinylated anti-matureBDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of proBDNF present in samples and protein standards. A proBDNF positive control (QC sample) is provided to assure consistent assay performance.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular human blood samples.
Lieferumfang: Each kit contains 1 or 2 × 96 pre-coated strip plates, protein standards, detection reagents, wash and sample buffers and detailed instructions.
Bitte beachten For research use only. Not for diagnostic or clinical purposes.