Mouse Isotype Control (Unconjugated)

Lieferant: Biosensis
M-1249-100
BSENM-1249-100EA 236 EUR
BSENM-1249-100
Mouse Isotype Control (Unconjugated)
Antikörper
X63 is a control mouse IgG antibody with no known reactivity. The antibody has been used successfully a negative control in westerns, all forms of IHC, IP and FACS

No staining has ever been identified with this immunoglobulin demonstrating its non-specific value as a control.
Application Information:
Recommended for use as a control for Western Blotting, immunohistochemistry and FACS at a concentration equal to that of the test antibody. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Control
Antigen: Unknown
Clonality: Monoclonal
Clone: X63
Conjugation: Unconjugated
Epitope: This control IgG has no known binding ability.
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR