Anti-DYKDDDDK Mouse Monoclonal Antibody [clone: BFtag01]
Lieferant: Biosensis
M-1301-100
BSENM-1301-100EA 360 EUR
BSENM-1301-100
Anti-DYKDDDDK Mouse Monoclonal Antibody [clone: BFtag01]
Antikörper
The D-tag system utilises a short hydrophilic peptide (DYKDDDDK) that is fused to either the N- or C-terminus of the protein of interest. It can be used in conjunction with other tags such as the 6X His tag.
Mouse monoclonal anti D-tag antibody recognises over-expressed proteins containing the D-tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH) and Immunoprecipitation (IP). Suggested dilutions: WB at 1:100, IS and IP at 1:20. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: BFtag01
Conjugation: Unconjugated
Epitope: DYKDDDDK
Host: Mouse
Isotype:
Reactivity:
Bestellen