Human synthetic Abeta1-42 oligomers peptide
Lieferant: Biosensis
PE-1750-1000
BSENPE-1750-1000EA 112 EUR
BSENPE-1750-1000
Human synthetic Abeta1-42 oligomers peptide
Proteine und Peptide
A proprietary preparation of human amyloid beta peptide (amino acids 1-42) that was initially monomerized by HFIP-treatment and then allowed to form oligomers by the procedure described in Youmans KL et al., 2012, followed by lyophilisation using Biosensis’ proprietary stabilization procedures.
The resulting oligomeric mixture has been specially designed to allow the formation of stable, oligomeric Aβ1-42 peptide, multimeric complexes or oligomers. The material is intended to be used as a stable and consistent standard or positive control for oligomeric ELISA assays, as well as other research applications. This product is supplied as 2 x 500 ng vials, each containing lyophilized Aβ oligomers. Note that the amount of provided oligomeric protein is based on the amount of monomeric Aβ used to form these oligomers. The precise formation, size and number of oligomers cannot be quantified by any known method.