Actinomycin D

Lieferant: BIO SOURCE

PHZ1024
BSOUPHZ1024EA 0 EUR
BSOUPHZ1024
Actinomycin D
Actinomycin D
Formel: C₆₂H₈₆N₁₂O₁₆
MDL: MFCD00005033
CAS-Nummer: 50-76-0
UN: 3462
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR