Cytochalasin D

Lieferant: BIO SOURCE

PHZ1063
BSOUPHZ1063EA 0 EUR
BSOUPHZ1063
Cytochalasin D
Cytochalasin D
Formel: C₃₀H₃₇NO₆
Schmelzpunkt: 259…260 °C
MDL: MFCD00077706
CAS-Nummer: 22144-77-0
EINECS: 244-804-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR